Learn More
Medchemexpress LLC Boc-D-2,3-diaminopropionic acid | 76387-70-7 | MFCD01632072 | ≥97.0% | 204.22 g/mol | C8H16N2O4 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0146925G
Encompass_Preferred
Boc-D-2,3-diaminopropionic acid is a Boc-protected D-2,3-diaminopropionic acid used as a protected amino acid building block for peptide and organic synthesis. It enables selective protection of the α-amino group in multi-step syntheses and is employed in the synthesis of NMDA receptor glycine-site agonists with GluN2 subunit specificity.
  • Boc-protected amino acid suitable for peptide and organic synthesis.
  • Facilitates selective protection of the α-amino group.
  • High purity as confirmed by NMR analysis.
  • Available in small laboratory package sizes for research use.
  • Useful for preparing NMDA receptor glycine-site agonists and related analogs.
Catalog No. 50-003-02124