Learn More
Medchemexpress LLC Boc-D-alaninol | 106391-86-0 | 175.23 | 10 G

Supplier: Medchemexpress LLC HYW00818510G
Boc-D-alaninol is a laboratory chemical primarily used in the manufacture of various substances. It is supplied as a white to off-white solid. This product has not been fully validated for medical applications and is for research use only.
- High purity: 99.89% (GC)
- Molecular weight: 175.23
- Optimal storage for powder: -20°C for 3 years, or 4°C for 2 years
- Optimal storage in solvent: -80°C for 6 months, or -20°C for 1 month
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.