Learn More
Medchemexpress LLC Boc-D-Nip-OH | 163438-09-3 | 229.28 | 5 G

Supplier: Medchemexpress LLC HYW0029005G
Boc-D-Nip-OH is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
- Biochemical reagent for life science research
- Organic compound
- Purity 98.0%
- Molecular weight 229.28
- Solid appearance
- White to off-white color
- Storage (powder): -20°C for 3 years, 4°C for 2 years
- Storage (in solvent): -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.