Learn More
Medchemexpress LLC Boc-D-norleucine | 55674-63-0 | MFCD00063168 | 98.0% | 231.29 | 1 G
SDP
Supplier:  Medchemexpress LLC HY41912B1G
Encompass_Preferred
Boc-D-norleucine is a leucine derivative utilized in peptide synthesis. This chemical is supplied as a colorless to light yellow liquid.
  • Suitable for peptide synthesis
  • High purity for reliable results
  • Stable storage conditions
  • Good solubility in various solvents
Catalog No. 50-004-03472