Learn More
Enzo Life Sciences Boc-D(OMe)-FMK (1mg). CAS: 187389-53-3
SDP
Supplier:  Enzo Life Sciences ALX260071M001
Encompass_Preferred
Irreversible, cell permeable broad-spectrum caspase inhibitor. Alternative name: Caspase family inhibitor (fluoromethylketone). Formula: C11H18FNO5. MW: 263.3. Purity: ≥95% (TLC). Appearance: White to off-white powder. Solubility: Soluble in DMSO or dimethyl formamide. Long Term Storage: -20°C. Use/Stability: Stock solutions should be divided into aliquots and stored at -20°C. For long term storage keep aliquots at -70°C. Handling: Keep cool and dry. After reconstitution, prepare aliquots and store at -80°C.
Catalog No. 50-200-8282