Learn More
Cayman Chemical Boc-D-Phe-Pro-OH 50mg
SDP
Supplier:  Cayman Chemical 3115150 MG
Encompass_Preferred
A synthetic intermediate; has been used in the synthesis of peptide thrombin inhibitors
Catalog No. 50-298-9577