Learn More
Medchemexpress LLC Boc-D-phenylalanyl-D-phenylalanine | 125511-90-2 | 99.6% | 412.48 g/mol | C23H28N2O5 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW2501401G
Encompass_Preferred
Boc-D-phenylalanyl-D-phenylalanine is a Boc-protected D-dipeptide used as a building block for peptide synthesis and biochemical research. It is supplied as a white to off-white solid with reported high purity, molecular formula C23H28N2O5, and molecular weight 412.48 g/mol. CAS number 125511-90-2. Typical laboratory pack sizes cover small-scale synthesis and assay use.
Catalog No. 50-002-80870