Learn More
Medchemexpress LLC Boc-Dap-NE | 160800-65-7 | 99.9% | C23H36N2O5 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY78931B
Encompass_Preferred
Boc-Dap-NE is an intermediate in the synthesis of Monomethyl auristatin E, which is an inhibitor of tubulin polymerization. Monomethyl auristatin E can be used to synthesize Antibody-Drug Conjugates (ADCs) as an ADC Cytotoxin.
  • Intermediate in the synthesis of Monomethyl auristatin E.
  • Inhibitor of tubulin polymerization.
  • Can be used to synthesize Antibody-Drug Conjugates (ADCs).
  • Purity of 99.94%.
Catalog No. 50-003-59225