Learn More
Medchemexpress LLC Boc-Dap-NE | 160800-65-7 | MFCD28385913 | >98.0% | 420.5 | C23H36N2O5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY7893110MM 1ML
Encompass_Preferred
Boc-Dap-NE is a protected diaminopropionic acid derivative used as an intermediate in the synthesis of monomethyl auristatin E (MMAE) and related antibody-drug conjugates. It is supplied as solid material and as a DMSO solution for small-scale research and synthesis applications.
  • Used as an intermediate in ADC and MMAE synthesis.
  • Available as ≥98% pure solid and as a 10 mM solution in DMSO.
  • Suitable for peptide coupling and linker assembly chemistry.
  • Supplied in small research quantities for medicinal chemistry workflows.
Catalog No. 50-002-04104