Learn More
Medchemexpress LLC Boc-Gln-Ala-Arg-AMC (acetate) | 00-00-0 | 99.7% | C31H46N8O10 | 10MG
SDP
Supplier:  Medchemexpress LLC HY134432B10MG
Encompass_Preferred
Boc-Gln-Ala-Arg-AMC acetate is a fluorogenic peptide substrate that releases 7-amino-4-methylcoumarin (AMC) upon proteolysis. It is used to quantify trypsin activity and has been applied for measuring TMPRSS2 proteolytic activity. The compound is supplied in research-scale quantities and is provided with safety and quality documentation.
  • Fluorogenic substrate releasing AMC upon proteolytic cleavage.
  • Suitable for measuring trypsin and TMPRSS2 activity in fluorescence assays.
  • Available in small research-scale quantities for assay development.
  • Accompanied by safety data sheet and certificate of analysis for quality assurance.
  • Soluble in common assay solvents such as DMSO for easy assay preparation.
Catalog No. 50-002-59286