Learn More
Medchemexpress LLC Boc-Glu-OBzl | 30924-93-7 | MFCD00065568 | 337.37 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW007875
Encompass_Preferred
Boc-Glu-OBzl is a glutamic acid derivative used as an N-terminal protected amino acid in solid-phase peptide synthesis (SPPS) to create unique peptides containing glutamate benzyl ester residues. It also serves as a substrate for Vitamin K-dependent carboxylation.
  • Influences anabolic hormone secretion
  • Supplies fuel during exercise
  • Improves mental performance under stress
  • Prevents exercise-induced muscle damage
  • Used in peptide synthesis
Catalog No. 50-004-49861