Learn More
Medchemexpress LLC Boc-Gly-Gly-Phe-Gly-OH | 187794-49-6 | 99.6% | 436.46 | 100 MG

Supplier: Medchemexpress LLC HYP1449100MG
Boc-Gly-Gly-Phe-Gly-OH is a self-assembly of N- and C-protected tetrapeptide, functioning as a protease cleavable linker for antibody-drug conjugates (ADC).
- Functions as a protease cleavable linker
- Self-assembly of N- and C-protected tetrapeptide
- White to off-white solid appearance
- Molecular formula: C20H28N4O7
- Soluble in DMSO (≥ 100 mg/mL) and H2O (25 mg/mL)
- Recommended storage for powder: -80°C for 2 years; -20°C for 1 year
- Recommended storage in solvent: -80°C for 6 months; -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.