Learn More
Medchemexpress LLC Boc-Gly-Gly-Phe-Gly-OH | 187794-49-6 | 99.62% | 436.46 | 1 G

Supplier: Medchemexpress LLC HYP14491G
Boc-Gly-Gly-Phe-Gly-OH is a self-assembly of N- and C-protected tetrapeptide, serving as a protease cleavable linker for antibody-drug conjugates (ADC). These conjugates are composed of an antibody to which a cytotoxin is attached via a linker.
- Used for antibody-drug conjugates
- Protease cleavable linker
- Target: ADC linker
- Appearance: White to off-white solid
- Soluble in DMSO and H2O
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.