Learn More
Medchemexpress LLC Boc-Gly-Gly-Phe-Gly-OH TFA | 2450273-39-7 | 98.3% | 550.48 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP1449A5MG
Encompass_Preferred
Boc-Gly-Gly-Phe-Gly-OH TFA is a self-assembly of N- and C-protected tetrapeptide. It acts as a protease cleavable linker, primarily used for antibody-drug conjugates (ADC).
  • Self-assembly of N- and C-protected tetrapeptide
  • Protease cleavable linker
  • Used for antibody-drug conjugates (ADC)
  • Suitable for cancer research
  • For research use only
Catalog No. 50-004-24397