Learn More
Medchemexpress LLC Boc-Gly-OMe | 31954-27-5 | 99.9% | 189.21 g/mol | C8H15NO4 | 500 G

Supplier: Medchemexpress LLC HYW041862500G
Boc-Gly-OMe is the tert-butyloxycarbonyl (Boc) protected methyl ester of glycine, used as an intermediate and building block in peptide and organic synthesis research.
- High purity (99.9%).
- Molecular weight 189.21 g/mol.
- Chemical formula C8H15NO4.
- Available in bulk package sizes, including 500 G.
- Intended for research and analytical applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.