Learn More
Bachem Boc-Ile-Glu-Gly-Arg-AMC
SDP
Supplier:  Bachem 4004961.0025
Encompass_Preferred
Boc-Ile-Glu-Gly-Arg-AMC Acetate, 65147-06-0, C34H50N8O10, Mr 730.82, 25mg. Boc-IEGR-AMC, a specific, highly fluorogenic substrate for clotting enzyme factor Xa (coagulation factor Xa). Boc-IEGR-AMC is also hydrolyzed by acrosin from the ascidian Halocynthia roretzi. The use of this substrate in the indirect determination of coagulation factor IX has been described.
Catalog No. 50-194-6934