Learn More
Medchemexpress LLC Boc-L-alanine benzyl ester | 51814-54-1 | MFCD00190775 | 95.0% | 279.33 g/mol | C15H21NO4 | 10 G

Supplier: Medchemexpress LLC HYW04186410G
Boc-L-alanine benzyl ester is a Boc-protected derivative of L-alanine used as a protected building block in peptide and organic synthesis.
- CAS number: 51814-54-1.
- Molecular formula: C15H21NO4.
- Molecular weight: 279.33 g/mol.
- Purity: 95.0%.
- Appearance: Colorless to off-white; solid below 25.5°C, liquid above 26°C.
- Storage: Pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
- Typical application: Protected amino acid for peptide and organic synthesis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.