Learn More
Medchemexpress LLC Boc-L-Arg(Mtr)-OH | 102185-38-6 | 99.1% | 486.58 | C21H34N4O7S | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW077219
Encompass_Preferred
Boc-L-Arg(Mtr)-OH is an N-Boc protected L-arginine derivative bearing an Mtr-protected guanidine side chain, supplied for use as a building block in peptide synthesis and related organic syntheses.
  • N-Boc protection on the α-amino group.
  • Mtr protecting group on the arginine guanidine side chain.
  • Suitable for solid-phase and solution-phase peptide synthesis.
  • High purity for research applications.
  • Available in multiple pack sizes, including 25 g.
Catalog No. 50-002-09019