Learn More
Bachem Boc-Lys-OMe
SDP
Supplier:  Bachem 4002809.0005
Encompass_Preferred
Boc-Lys-OMe Acetate, 55757-60-3, C12H24N2O4, Mr 260.33, 5g. Boc-L-lysine methyl ester is efficiently hydrolyzed by thrombin and especially trypsin.
Catalog No. 50-194-7078