Learn More
STA PHARMACEUTICAL US LLC Boc-L-Orn(Fmoc)-OH | 25 g | CAS 150828-96-9 | MDL MFCD00080279
Supplier: STA PHARMACEUTICAL US LLC STAAA1153025GM
Boc-L-Orn(Fmoc)-OH is a Amino Acid reagent (Subcategory: Lys) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 150828-96-9
- MDL: MFCD00080279
- InChIKey: YEBWACZYMHWWEK-NRFANRHFSA-N
- Molecular Weight: 454.523
- Molecular Formula: C25H30N2O6
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (S)-5-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-((tert-butoxycarbonyl)amino)pentanoic acid
- SMILES: CC(OC(N[C@@H](CCCNC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O)C(O)=O)=O)(C)C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.