Learn More
Medchemexpress LLC Boc-L-phenylalanine | 13734-34-4 | MFCD00002663 | 99.8% | C14H19NO4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY7909610MM/1ML
Encompass_Preferred
Boc-L-phenylalanine is a Boc-protected L-phenylalanine that serves as a versatile building block in organic synthesis. It is utilized in the creation of other active compounds and functions effectively as a capping agent.
  • Used in the synthesis of other active compounds
  • Acts as a capping agent
  • Reacts completely with hydroxyl-terminated PLLA
  • Enables generation of amino-terminated PLLA
  • Applicable for block copolymer synthesis
Catalog No. 50-004-34905