Learn More
Medchemexpress LLC Boc-Leu-Gly-Arg-AMC | 65147-09-3 | 98.3% | 601.69 | 10 MG

Supplier: Medchemexpress LLC HYP223710MG
Boc-Leu-Gly-Arg-AMC is a fluorogenic AMC substrate for the convertases and can be used in enzymatic assays.
- Fluorogenic AMC substrate for convertases
- Can be used in enzymatic assays
- Optimal activity at pH 9.0 and 45 °C for cleaving Boc-Leu-Gly-Arg-AMC
- Molecular formula: C29H43N7O7
- Sequence: Boc-Leu-Gly-Arg-AMC
- Sequence shortening: Boc-LGR-AMC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.