Learn More
Carbosynth LLC. Boc-Leu-Gly-Arg-MCA (5 mg vial), 1ea, MW: 601.69, Molecular formula: C29H43N7O7, CAS# 65147-09-3
SDP
Supplier:  Carbosynth LLC. MLG3102V5MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: t-Butyloxycarbonyl-L-Leucyl-Glycyl-L-Arginine 4-Methyl-Coumaryl-7-Amide, Boc-Leu-Gly-Arg-AMC, Boc-LGR-AMC, Boc-LGR-MCA, Purity: , Grade AA, Application: Substrate for Horseshoe Crab Clotting Enzyme S, Salt Form:, Hydrochloride Form, Storage Temperature: -20C, References: S. Iwanaga, T. Morita, T.Harada, S. Nakamura, M. Niwa, K. Takada, T. Kimura and S. Sakakibara, Haemostasis, 7, 183 (1978).
Catalog No. 50-168-5987