Learn More
Medchemexpress LLC Boc-Leu-Lys-Arg-AMC | 109358-47-6 | 96.91% | 672.82 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP43125MG
Encompass_Preferred
Boc-Leu-Lys-Arg-AMC is a fluorogenic substrate for the Kex2 endoprotease, intended for research use only.
  • Fluorogenic substrate for Kex2 endoprotease
  • For research use only
Catalog No. 50-003-97643