Learn More
Apexbio Technology LLC Boc-N-Me-Leu-OH 53363-89-6 5g
SDP
Supplier:  Apexbio Technology LLC A66515000
Encompass_Preferred
Boc-N-Me-Leu-OH (53363-89-6) is an N-methylated leucine derivative containing a tert-butoxycarbonyl (Boc) protecting group It is employed in peptide synthesis to modify peptide secondary structure and explore structural constraints in peptide systems Based on these properties Boc-N-Me-Leu-OH holds research potential in investigating biomolecular interactions enzyme substrate specificity receptor binding affinity and in the preparation of peptides for studying conformation-dependent activity toward proteins enzymes and receptors
Catalog No. 50-224-0406