Learn More
Medchemexpress LLC Boc-N-methyl-L-isoleucine | 52498-32-5 | 98.7% | 245.32 | 5 G

Supplier: Medchemexpress LLC HYW0120005G
Boc-N-methyl-L-isoleucine (Boc-N-Me-Ile-OH) is a peptide product that can be used as a precursor in organic synthesis and pharmaceuticals.
- Purity: 98.70%
- Molecular weight: 245.32
- Formula: C12H23NO4
- Appearance: Solid
- Color: White to off-white
- Solubility (In Vitro) in DMSO: 40 mg/mL (163.05 mM; need ultrasonic)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.