Learn More
Medchemexpress LLC Boc-N-PEG1-C2-NHS ester | 1260092-55-4 | 95.0% | C14H22N2O7 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY141116100MG
Encompass_Preferred
Boc-N-PEG1-C2-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Utilizes the ubiquitin-proteasome system
  • Enables selective degradation of target proteins
Catalog No. 50-003-55565