Learn More
Medchemexpress LLC Boc-N-PEG1-C2-NHS ester | 1260092-55-4 | 97.0% | C14H22N2O7 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY141116
Encompass_Preferred
Boc-N-PEG1-C2-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs are designed with two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based linker
  • Used in the synthesis of PROTACs
  • Selectively degrades target proteins
  • Utilizes the intracellular ubiquitin-proteasome system
Catalog No. 50-003-52602