Learn More
Medchemexpress LLC Boc-N-PEG1-C2-NHS ester | 1260092-55-4 | 97.0% | C14H22N2O7 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY141116500MG
Encompass_Preferred
Boc-N-PEG1-C2-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. They consist of two different ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other targeting a specific protein.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system
  • For selective degradation of target proteins
  • Consists of two distinct ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • Other ligand targets a specific protein
Catalog No. 50-003-55628