Learn More
Medchemexpress LLC Boc-NH-PEG-amine | ≥98.0% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY140727
Encompass_Preferred
Boc-NH-PEG-amine (MW 5000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase, and the other targets a specific protein. These compounds leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Average molecular weight of 5000
  • White to off-white solid
  • Purity of ≥98.0%
  • Functions as a PEG-based PROTAC linker
  • Applicable in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system for protein degradation
  • For research use only
Catalog No. 50-003-52580