Learn More
Medchemexpress LLC Boc-NH-PEG10-NHS ester | 3075546-11-8 | 98.0% | C32H58N2O16 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY141120100MG
Encompass_Preferred
Boc-NH-PEG10-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are molecules with two ligands connected by a linker; one targets an E3 ubiquitin ligase and the other targets a protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • Utilizes the intracellular ubiquitin-proteasome system
  • For research use only
Catalog No. 50-003-55622