Learn More
Medchemexpress LLC Boc-NH-PEG10-NHS ester | 98.0% | C32H58N2O16 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14112050MG
Encompass_Preferred
Boc-NH-PEG10-NHS ester is a PEG-based PROTAC linker for the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting two ligands: one targeting an E3 ubiquitin ligase and the other the protein of interest. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Leverages intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Composed of two ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • The other ligand targets the protein of interest
Catalog No. 50-003-55132