Learn More
Medchemexpress LLC Boc-NH-PEG15-NH2 | 2222566-55-2 | 98.0% | C37H76N2O17 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140235100MG
Encompass_Preferred
Boc-NH-PEG15-NH2 is a chemical compound offered by MedChemExpress, identified by CAS number 2222566-55-2 and chemical formula C37H76N2O17. This product is an off-white oil with a purity of 98.0% by NMR. It is stable under recommended conditions and classified as not hazardous, making it suitable for laboratory use and substance manufacturing.
  • Colorless to off-white oil
  • 98.0% purity by NMR
  • Store at -20°C, sealed and away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
  • Shipped at room temperature if less than 2 weeks
  • Suitable for laboratory chemical applications
  • Stable under recommended storage conditions
  • Avoid contact with strong acids/alkalis and oxidizing/reducing agents
Catalog No. 50-003-53511