Learn More
Medchemexpress LLC Boc-NH-PEG2-C2-NH2 | 153086-78-3 | 248.32 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00847410G
Encompass_Preferred
Boc-NH-PEG2-C2-NH2 (PROTAC Linker 13) is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs are molecules that utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins, featuring two ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Designed for selective degradation of target proteins
  • For research use only
Catalog No. 50-004-47153