Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-nh-peg2-ch2ch2cooh | 1365655-91-9 | MFCD22056304 | 98.0% | 277.31 g/mol | C12H23NO6 | 1 G
SDP

Supplier:  Medchemexpress LLC HYW0258121G

Encompass_Preferred

Boc-NH-PEG2-CH2CH2COOH is a short PEG2-based linker with a Boc-protected terminal amine and a terminal carboxylic acid, designed for use in PROTAC synthesis and other bioconjugation applications. It offers a compact, hydrophilic spacer to connect ligands or warheads while maintaining solubility and providing reliable coupling chemistry for assembly of bifunctional molecules.

  • Provides Boc-protected terminal amine for selective deprotection
  • Contains a short PEG2 spacer to improve flexibility and solubility
  • Features a terminal carboxylic acid for standard amide coupling
  • Supplied at high purity (~98%) for research use
  • Soluble in DMSO with established in vivo formulation protocols
  • Available in multiple laboratory pack sizes

Catalog No. 50-003-01459


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.