Learn More
Medchemexpress LLC HY-W005056 100mg Medchemexpress, Boc-NH-PEG2-CH2COOH CAS:108466-89-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW005056100MG
Encompass_Preferred
Medchemexpress, HY-W005056 100mg Boc-NH-PEG2-CH2COOH CAS:108466-89-3 Boc-NH-PEG2-CH2COOH is a PEG-based PROTAC linker can be used in the synthesis of PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3226
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.