Learn More
Medchemexpress LLC Boc-NH-PEG2-NH-Boc | 475591-59-4 | ≥97.0% | C16H32N2O6 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1411981G
Encompass_Preferred
Boc-NH-PEG2-NH-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-55682