Learn More
Medchemexpress LLC Boc-NH-PEG2-NH-Boc | 475591-59-4 | ≥97.0% | C16H32N2O6 | 500 MG
Supplier: Medchemexpress LLC HY141198500MG
Boc-NH-PEG2-NH-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
- Used as a PROTAC linker for synthesizing PROTACs
- Utilized to selectively degrade target proteins
- Leverages the intracellular ubiquitin-proteasome system