Learn More
Medchemexpress LLC Boc-NH-PEG24-CH2CH2COOH | 2839118-68-0 | 96.0% | C56H111NO28 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14047450MG
Encompass_Preferred
This PEG-based PROTAC linker is used in the synthesis of PROTACs. These compounds are designed with two distinct ligands joined by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets a specific protein. PROTACs leverage the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-57027