Learn More
Medchemexpress LLC Boc-NH-PEG3-CH2COOH | 462100-06-7 | 97.0% | 307.34 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY33366100MG
Encompass_Preferred
This product is a PEG-based PROTAC linker, which can be utilized in the synthesis of PROTACs. PROTACs are molecules designed to exploit the intracellular ubiquitin-proteasome system, leading to the selective degradation of target proteins.
  • Used in the synthesis of PROTACs.
  • Facilitates targeted protein degradation.
  • Leverages the ubiquitin-proteasome system.
  • Available in various quantities.
  • Purity of 97.0%.
Catalog No. 50-004-04692