Learn More
Cayman Chemical Boc-NH-PEG3-OH 500mg
SDP
Supplier:  Cayman Chemical 30496500 MG
Encompass_Preferred
A PEGylated building block; has been used as a spacer in biotinylated heterobifunctional cross-linker agents; has been used as a spacer in near infrared imaging probes for CYP1B1, which is expressed in a variety of tumors; has also been used in the synthesis of graftable peptidomimetics that bind to human αVβ3 integrin and inhibit adhesion of human osteoprogenitor cells to vitronectin
Catalog No. 50-299-0639