Learn More
Medchemexpress LLC Boc-NH-PEG4-C2-Boc | 2138484-13-4 | 421.53 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13837050MG
Encompass_Preferred
Boc-NH-PEG4-C2-Boc is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. This compound facilitates the development of molecules that leverage the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins. It is intended for research use only.
  • PEG-based linker structure
  • Essential for PROTAC synthesis
  • Enables targeted protein degradation via ubiquitin-proteasome system
Catalog No. 50-003-49468