Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-NH-PEG4-CH2COOH | 876345-13-0 | MFCD30528568 | ≥97.0% | 351.39 g/mol | C15H29NO8 | 5 G
SDP

Supplier:  Medchemexpress LLC HY426405G

Encompass_Preferred

Boc-NH-PEG4-CH2COOH is a Boc-protected polyethylene glycol (PEG4) linker with a terminal carboxylic acid group used as a building block in bioconjugation chemistry, including PROTACs, antibody-drug conjugates, and peptide coupling.

  • Boc-protected primary amine enabling orthogonal deprotection
  • PEG4 spacer provides solubility and flexibility
  • Terminal carboxylic acid suitable for amide coupling
  • Molecular weight 351.39 g/mol
  • High purity ≥97.0%
  • CAS number 876345-13-0 for unambiguous identification

Catalog No. 50-002-03205


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.