Learn More
Medchemexpress LLC Boc-NH-PEG4-MS | 1447797-72-9 | 98.0% | C14H29NO8S | 500 MG
SDP
Supplier:  Medchemexpress LLC HY140391500MG
Encompass_Preferred
Boc-NH-PEG4-MS is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These molecules are designed to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. This product is intended for research use only and has not been fully validated for medical applications.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Exploits the ubiquitin-proteasome system
  • Aids in selective degradation of target proteins
  • Available as a colorless to light yellow liquid
  • Stable under recommended storage conditions
  • For research use only
Catalog No. 50-003-56928