Learn More
Medchemexpress LLC Boc-NH-PEG4-NHS ester | 859230-20-9 | 99.1% | C20H34N2O10 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY141119
Encompass_Preferred
Boc-NH-PEG4-NHS ester is a PEG-based PROTAC linker designed for the synthesis of PROTACs. These PROTACs operate by connecting two ligands: one targets an E3 ubiquitin ligase, and the other targets a specific protein. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Utilized for PROTAC synthesis
  • Exploits the ubiquitin-proteasome system
  • Enables selective degradation of target proteins
Catalog No. 50-003-52590