Learn More
Medchemexpress LLC Boc-NH-PEG6-amine | 1091627-77-8 | 97.0% | C19H40N2O8 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1402315G
Encompass_Preferred
Boc-NH-PEG6-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds that contain two distinct ligands linked together: one that binds to an E3 ubiquitin ligase and another that targets a protein of interest. These molecules work by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-57090