Learn More
Medchemexpress LLC Boc-nh-peg6-amine | 1091627-77-8 | MFCD00676451 | ≥98% | C18H38N2O7 | 10 G
SDP
Supplier:  Medchemexpress LLC HY14023110G
Encompass_Preferred
Boc-NH-PEG6-amine is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Enables the synthesis of PROTACs with improved solubility and bioavailability
  • Provides a flexible and biocompatible linker for PROTACs
Catalog No. 50-003-53506