Learn More
Medchemexpress LLC Boc-NH-PEG6-CH2CH2COOH | 882847-13-4 | MFCD07781255 | 97.0% | 453.52 | C20H39NO10 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0402441G
Encompass_Preferred
Boc-NH-PEG6-CH2CH2COOH is a Boc-protected PEG6 linker with a terminal carboxylic acid used as a building block in PROTAC, ADC, and conjugation chemistry. It provides a hydrophilic PEG spacer and a Boc-protected amine for orthogonal functionalization.
  • Boc-protected amine for orthogonal functionalization.
  • PEG6 spacer imparts hydrophilicity and flexibility.
  • Terminal carboxylic acid enables standard amide coupling.
  • High purity suitable for research applications.
  • Available in small-scale quantities for synthesis workflows.
Catalog No. 50-002-02900