Learn More
Medchemexpress LLC Boc-nh-peg7-propargyl | 2112737-90-1 | C22H41NO9 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1408821G
Encompass_Preferred
Boc-NH-PEG7-propargyl is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that possess Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAC) with azide groups
Catalog No. 50-003-55407