Learn More
Medchemexpress LLC Boc-NH-PEG7-propargyl | 2112737-90-1 | C22H41NO9 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140882
Encompass_Preferred
Boc-NH-PEG7-propargyl is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups. This compound is instrumental in constructing PROTACs, which are designed to selectively degrade target proteins by exploiting the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules
  • Exploits the intracellular ubiquitin-proteasome system for target protein degradation
Catalog No. 50-003-52567